missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plant Genomic DNA Purification Kit

Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types.

Brand:  Thermo Scientific™ K0791

99.45 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 11571645

  • £153.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

Thermo Scientific GeneJET Plant Genomic DNA Purification Mini Kit is designed for rapid and efficient purification of high quality genomic DNA from a wide variety of plant species and tissue types. It utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.

Highlights
Fast—procedure completed in 30 min following cell lysis
Convenient—protocol does not include clogging-prone lysate filtration step
High yield—up to 32 μg of high quality DNA from 100 mg sample, including nuclear, mitochondrial, and chloroplast DNA
Compatible—use with awide variety of samples (leaves, roots, sprouts and other parts of various plants)

The standard procedure takes less than 30 minutes following cell lysis and yields purified DNA greater than 30 kb in size. DNA yields vary between different species and tissues depending on genome size, ploidy, cell number, and the age of the tissue sample. The typical yield from a standard tissue source (e. g. young wheat leaves) is 30 to 32 μg from 100 mg of tissue. Isolated DNA can be used directly in PCR, qPCR, southern blotting, and enzymatic reactions.

Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Spin Column
Spin Column
Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, PCR
GeneJET
50 Preps
<20 min.
Spin Column
Plants
Not High-throughput Compatible (Manual)
Genomic DNA Purification Kit
Genomic DNA
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GeneJET Plant Genomic DNA Purification Kit

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Guanidine hydrochloride, Sodium lauryl sulfate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.