missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plant RNA Purification Kit

GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

Brand:  Thermo Scientific™ K0802

811.50 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11501655

  • £1082.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Dithiothreitol (DTT, #R0861)
This item is not returnable. View return policy

Description

Description

GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample. See Table 1 for typical total RNA yields from various sources.

Thermo Scientific GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample. See Table 1 for typical total RNA yields from various sources.

Highlights
Fast—procedure completed in 20 min following cell lysis
Convinient—protocol does not include clogging-prone lysate filtration step
High yield—up to 65 μg of high purity and high integrity RNA from 50 mg plant sample
Compatible with wide variety of samples (leaves, roots, sprouts and other parts of various plants)

Applications
Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:
• RT-PCR
• RT-qPCR
• Northern blotting
• Nuclease protection assay

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 200 mL Plant RNA Lysis Solution; store at room temperature
• 200 mL Wash Buffer WB 1 (concentrated); room temperature
• 2 x 100 mL Wash Buffer 2 (concentrated); room temperature
• 125 mL Water, Nuclease-free; room temperature
• 250 GeneJET RNA Spin Columns with Collection Tubes; room temperature
• 250 Collection Tubes (2 mL); room temperature
• 250 Collection Tubes (1.5 mL); room temperature
20 min. (After sample homogenization)
Plants
Not High-throughput Compatible (Manual)
250 Preps
Room Temperature
Up to 65 μg
Silica Spin Column
50 μL
Total RNA
RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Mini
≤100 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.