missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plasmid Miniprep Kit

Product Code. p-4530580 Shop All Thermo Scientific Products
Click to view available options
Quantity:
250 Preps
50 Preps
Unit Size:
250 reactions
75mL
Each
This item is not returnable. View return policy

Product Code. 10319699

Brand: Thermo Scientific™ K0502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column.

Thermo Scientific GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column. The kit recovers up to 20 μg of high copy plasmid DNA per isolation procedure.

Highlights

  • Efficient—high yields of up to 20 μg of high quality plasmid DNA
  • Fast—procedure takes less than 14 minutes
  • Convenient—no phenol/chloroform extraction or alcohol precipitation required
  • Pure—purified DNA is immediately ready to use

Applications

Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:

  • FastDigest or conventional restriction digestion
  • Automated fluorescent and radioactive sequencing
  • PCR
  • In vitro transcription
  • Transformation

Principle

A bacterial culture is harvested and lysed. The lysate is then cleared by centrifugation and applied on the silica column to selectively bind DNA molecules at a high salt concentration. The adsorbed DNA is washed to remove contaminants, and the pure plasmid DNA is eluted in a small volume of elution buffer or water. The purified DNA is ready for immediate use in all molecular biology procedures, such as fast and conventional digestion with restriction enzymes, PCR, in vitro transcription, transformation, and automated sequencing.

Components

  • Resuspension Solution
  • Lysis Solution
  • Neutralization Solution Wash Solution (concentrated)
  • RNase A Solution
  • Elution Buffer
  • Spin Columns
  • Collection Tubes
  • Detailed Protocol

Related Products

  • RNase A (Part No.: R1253)
  • Elution Buffer for GeneJET Plasmid Miniprep Kit (Part No.: R1263)
  • Lysis Solution for GeneJET Plasmid Miniprep Kit (Part No.: R1223)
  • Resuspension Solution for GeneJET Plasmid Miniprep Kit (Part No.: R1213)
  • Neutralization Solution for GeneJET Plasmid Miniprep Kit (Part No.: R1233)
  • Wash Solution for GeneJET Plasmid Miniprep Kit (Part No.: R1243)
  • Binding Buffer for GeneJET Plasmid Miniprep Kit(Part No.: R1331)
TRUSTED_SUSTAINABILITY

Specifications

Column Type Spin Column
Format Spin Column
Isolation Technology Silica Column
Scale Mini
Sample Type Bacteria
For Use With (Application) PCR, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, In Vitro Transcription
High-throughput Compatibility Compatible (Vacuum manifold)
Prep Scale < 100 μg (Small-Scale) Plasmid DNA
Product Line GeneJET
Product Type Plasmid MiniPrep Kit
Quantity 50 Preps
Target Plasmid DNA
Test Time <14 min
Show More Show Less
compliance-icons
Product Identifier
  • GeneJET Plasmid Miniprep Kit
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.