Learn More
Thermo Scientific™ GeneJET RNA Cleanup and Concentration Micro Kit
GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.
£177.00 - £696.00
Specifications
Format | Spin Column |
---|---|
Isolation Technology | Spin Column |
Purification Time | 5 min. |
Sample Type | RNA |
High-throughput Compatibility | Not High-throughput Compatible (Manual) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13203159
|
Thermo Scientific™
K0841 |
50 Preps |
£177.00
50g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13213159
|
Thermo Scientific™
K0842 |
250 Preps |
£696.00
250 tests |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.
The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.
Thermo Scientific GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.
The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.
Highlights
• Advanced column design—easy to open, load and handle.
• Elute in 6 to 10 μL volume.
• 85 to 100% recovery of a high integrity RNA.
• Fast and simple—procedure takes less than 5 minutes.
Applications:
• The purified RNA can be used in common downstream applications
• RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis
Specifications
Spin Column | |
5 min. | |
Not High-throughput Compatible (Manual) | |
GeneJET | |
Total RNA |
Spin Column | |
RNA | |
Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Nucleic Acid Labeling, Northern Blotting, cDNA Library Construction | |
Micro |
For Research Use Only. Not for use in diagnostic procedures.