Learn More
Thermo Scientific™ GeneRuler 100 bp DNA Ladder
GREENER_CHOICE
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.
Brand:  Thermo Scientific™ SM0241
50.83 GBP valid until 2025-02-28
Use promo code "24154" to get your promotional price.
 View more versions of this product
Product Code. 10588170
  • £54.50 / 50µg
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
GeneRuler 100bp 100 to 1000bpDNA Ladder is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler 100bp DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).
The ladder is supplied with 6X DNA Loading Dye.
The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 100 bp DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase, protocol. The ladder is supplied with 6X DNA Loading Dye.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
No
Agarose Gels
GeneRuler
Agarose
DNA Ladder
100 to 1000 bp
Sustainable packaging
50 μg