missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneRuler 100 bp Plus DNA Ladder
GREENER_CHOICE

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.

£57.00 - £226.00

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Product Type DNA Ladder
Ready to Load No
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10309339
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
SM0321
50 μg
£57.00
50µg
In Stock
to see stock.
Add to basket
 
11853963
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
SM0322
5 x 50 μg
£226.00
Pack of 5
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

GeneRuler 100bp Plus DNA Ladder 100 to 3000bp is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler 100bp Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).

The ladder is supplied with 6X DNA Loading Dye.

Highlights

  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA
Specifications

Specifications

6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
No
Agarose Gels
GeneRuler
Agarose
DNA Ladder
100 to 3000 bp
Sustainable packaging
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.