Learn More
Thermo Scientific™ GeneRuler Ultra Low Range DNA Ladder

Perform sizing and approximate quantification of small dsDNA fragments on agarose or polyacrylamide gels. Ideal for siRNA analysis.
£70.75 - £267.00
Specifications
Concentration | 6X |
---|---|
Gel Type | Agarose |
Label or Dye | Xylene Cyanol FF, Bromophenol Blue, Orange G |
Product Type | DNA Ladder |
Ready to Load | No |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10150750
![]() |
Thermo Scientific™
SM1211 |
50 μg |
£70.75
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11823983
![]() |
Thermo Scientific™
SM1212 |
5 x 50 μg |
£267.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GeneRuler Ultra Low Range DNA Ladder is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. It is ideal for denaturing polyacrylamide gel electrophoresis. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler Ultra Low Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase.
Highlights
- Ideal for both DNA sizing and approximate quantification
- Ideal for non-denaturing polyacrylamide gel electrophoresis
- Ideal for analysis of siRNA
- Sharp bands
- Bright reference bands (given in red)
- Supplied with loading dye for sample DNA
Specifications
6X | |
Xylene Cyanol FF, Bromophenol Blue, Orange G | |
No | |
Agarose Gels | |
GeneRuler |
Agarose | |
DNA Ladder | |
10 to 300 bp | |
Sustainable packaging |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.