Learn More
Thermo Scientific™ Lambda DNA
Generate DNA size marker fragments with this linear, double-stranded DNA, a common substrate for restriction endonucleases.
Brand: Thermo Scientific™ SD0011
57.58 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 88
Additional Details : Weight : 0.00100kg
Description
Lambda is a temperate Escherichia coli bacteriophage. The virion DNA is linear and double-stranded (48502 nt) with 12 bp single-stranded complementary 5'-ends. After the phage particle injects its chromosome into the cell, the chromosome circularizes by end joining. In the lytic pathway, phage genes encoding replication, lysis, and virion proteins are expressed. The chromosome replicates, and the replicas are cleaved and packaged into progeny phage particles. In the lysogenic pathway, phage gene expression is repressed, and the circular chromosome inserts into the bacterial chromosome by recombination.
Highlights
Phage lambda DNA is a common substrate for restriction endonucleases and for generating DNA size marker fragments. For large scale isolation of phage DNA, cI857Sam7, a mutant carrying four known mutations, is often used. The DNA sequence used to construct a phage alpha restriction map includes these mutations. The map shows enzymes that cut lambda DNA once. Enzymes produced by Thermo Scientific are shown in orange (see figure bellow).
Specifications
0.3 μg/μL | |
DNA | |
[other] | |
500 μg |
DNA | |
Cloning/Nucleic Acid Gel Electrophoresis | |
Lambda DNA | |
Lambda phage |
For Research Use Only. Not for use in diagnostic procedures.