missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ M13/pUC Reverse Sequencing Primer (-26), 17-mer

Thermo Scientific M13/pUC Reverse Sequencing Primer (-26), 17-mer is a synthetic single-stranded 17-mere oligonucleotide with free 5'- and 3'-hydroxyl ends.

Brand:  Thermo Scientific™ SO101

20.70 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10754311

  • £41.40 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific M13/pUC Reverse Sequencing Primer (-26), 17-mer is a synthetic single-stranded 17-mere oligonucleotide with free 5'- and 3'-hydroxyl ends. This M13/pUC primer anneals to the region in the 5'-terminus of the lacZ gene. All primers are supplied as 10 μM aqueous solutions.

Applications
• Sequencing of DNA fragments inserted into the MCS within the lacZ gene of various cloning vectors, such as pUC19 (Cat. No. SM0221), pTZ19R, pTZ57R, M13mp18, and pBluescript II
• Colony screening by PCR

Primer Sequence: 5'-d(CAGGAAACAGCTATGAC)-3'

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sequencing Primer
Dry Ice
M13
pUC19, pTZ19R, pTZ57R, M13mp18, pBluescript II
Liquid
M13/pUC Reverse Sequencing Primer (-26), 17-mer, 10 μM (60 μL)

Store at –20°C.
10 μM
60 μL
Sequencing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.