Learn More
Thermo Scientific™ MassRuler DNA Ladder Mix, ready-to-use
GREENER_CHOICE
Perform acurate quantification and sizing of DNA fragments by agarose gel electrophoresis with this mixture of individual chromography-purified DNA fragments.
Brand:  Thermo Scientific™ SM0403
79.06 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10170690
  • £118.00 / Pack of 2
Estimated Shipment: 07-01-2025
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
MassRuler DNA Ladder Mix is specially designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. The intensity of the fragment in each ladder is calibrated against a standard that guarantees the precise quantity of each band. The ladders are mixtures of chromatography-purified, individual DNA fragments.
Highlights
  • Accurate DNA sizing and quantification -DNA ladder fragments are quantified spectrophotometrically
  • Sharp bands
  • Easy-to-remember fragment sizes and quantities
  • Ready-to-use -premixed with 6X MassRuler DNA Loading Dye for direct loading and room temperature storage
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6X
Bromophenol Blue
Yes
Agarose Gels
MassRuler
Agarose
DNA Ladder Mix
80 to 10000 bp
Sustainable packaging
2 x 500 μL