Learn More
Thermo Scientific™ MassRuler Express LR Forward DNA Ladder, ready-to-use
GREENER_CHOICE
Perform fast and reliable DNA quantification in short separation distances under various electrophoresis conditions with these specially designed DNA Ladder mixes.
Brand:  Thermo Scientific™ SM1263
Product Code. 10736041
  • £114.00 / Pack of 2
Explore more special offers
Description
Description
MassRuler Express LR Forward and Reverse DNA ladders are specially designed for fast and reliable DNA quantification in short separation distances under various electrophoresis conditions. In the forward versions, the mass of each DNA fragment is directly proportional to the fragment size, whereas in the reverse versions, the mass is inversely proportional to fragment's size.
The intensity of the fragment in each ladder is calibrated against a standard that guarantees the precise quantity of each band. The ladders are mixtures of chromatography-purified, individual DNA fragments.
Highlights
  • Accurate DNA sizing and quantification -DNA ladder fragments are quantified spectrophotometrically
  • Fast and reliable DNA quantification -short separation distances under various electrophoresis conditions
  • Sharp bands
  • Easy-to-remember fragment sizes and quantities
  • Ready-to-use -premixed with 6X MassRuler DNA Loading Dye for direct loading and room temperature storage
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6X
Bromophenol Blue
Yes
Agarose Gels
MassRuler
Agarose
DNA Ladder
100 to 1000 bp
Sustainable packaging
2 x 500 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3010542 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberSM1263
Lot Number2960122 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberSM1263
Lot Number3010766 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberSM1263
Lot Number3055695 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberSM1263
Lot Number3055684 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberSM1263
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ MassRuler Express LR Forward DNA Ladder, ready-to-use > 100 to 1000bp, 28 ng/μL