Learn More
Thermo Scientific™ Maxima Probe/ROX qPCR Master Mix (2X)
Optimize qPCR with probe chemistry using standard cycling protocols with these ready-to-use qPCR master mixes.
£188.00 - £2715.00
Specifications
Detection Method | Primer-probe |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11803923
|
Thermo Scientific™
K0231 |
200 reactions |
£188.00
200 reactions |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11813923
|
Thermo Scientific™
K0232 |
1,000 Reactions |
£880.00
1000 reactions |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11823923
|
Thermo Scientific™
K0233 |
4,000 Reactions |
£2715.00
4000 reactions |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.
Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.
Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.
Highlights
- Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity - detects low copy number targets
- Wide linear range - accurate quantification across 9 orders of magnitude
- Reproducibility and convenience - ready-to-use 2X master mix
Specifications
Primer-probe | |
High | |
Taq DNA Polymerase | |
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System | |
qPCR Master Mix | |
2X |
Tube | |
qPCR | |
Standard | |
Maxima | |
Gene Expression | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.