Learn More
Maxima Maxima SYBR Green qPCR Master Mix (2X), with separate ROX vial
Ready-to-use solution optimized for qPCR and 2-step RT-qPCR.
£205.00 - £3050.00
Specifications
Detection Method | SYBR |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11853923
|
Thermo Scientific™
K0251 |
200 reactions |
£205.00
200 reactions |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11340760
|
Thermo Scientific™
K0253 |
4,000 Reactions |
£3050.00
4000 reactions |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11863923
|
Thermo Scientific™
K0252 |
1,000 Reactions |
£946.00
1000 reactions |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.
Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.
- Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity - detects low copy number targets
- Wide linear range - accurate quantification across 9 orders of magnitude
- Reproducibility and convenience - ready-to-use 2X master mix
Specifications
SYBR | |
High | |
Taq DNA Polymerase | |
BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene | |
Real Time PCR SYBR Master Mix | |
2X |
Tube | |
qPCR | |
Standard | |
Maxima | |
Gene Expression | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.