Learn More
Maxima Maxima SYBR Green/Fluorescein qPCR Master Mix (2X)
Ready-to-use solution optimized for qPCR and 2-step RT-qPCR.
Brand: Thermo Scientific™ K0243
2308.60 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : A0
Additional Details : Weight : 0.19000kg
Includes:
- Maxima SYBR Green/ROX qPCR Master Mix (2x) is supplemented with ROX passive reference dye
- Maxima SYBR Green/Fluorescein qPCR Master Mix (2x) is supplemented with fluorescein passive reference dye
- Maxima SYBR Green qPCR Master Mix, ROX Solution provided (2x), comes with a separate ROX vial
- All Maxima SYBR Green qPCR Master Mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer. Supplied with nuclease-free water
Description
The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.
Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.
- Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity - detects low copy number targets
- Wide linear range - accurate quantification across 9 orders of magnitude
- Reproducibility and convenience - ready-to-use 2X master mix
Order Info
Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.
Specifications
Contains: • 4 x 12.5 mL Maxima SYBR Green/Fluorescein qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase, dNTPs (also dUTP) and SYBR Green I in an optimized PCR buffer with fluorescein passive reference dye; sufficient for 4000 x 25 μL reactions. • 2 x 30 mL Nuclease-Free Water Store at -20°C. |
|
Tube | |
qPCR | |
Standard | |
Maxima | |
4000 Reactions of 25 μL | |
2X | |
DNA, dsDNA |
SYBR | |
High | |
Taq DNA Polymerase | |
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5 | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
4,000 Reactions | |
4 x 12.5 mL |
For Research Use Only. Not for use in diagnostic procedures.