Learn More
Thermo Scientific™ MluI (10 U/μL)
The MluI restriction enzyme recognizes A^CGCGT sites and cuts best at 37°C in R buffer.
£91.25 - £434.00
Specifications
Concentration | 10 U/μL |
---|---|
Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
Enzyme | Mlu I |
Compatible Buffer | 10X Buffer R |
Sensitive to Heat Inactivation | Yes |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10540651
|
Thermo Scientific™
ER0561 |
1,000 units |
£91.25
1000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10141360
|
Thermo Scientific™
ER0562 |
5,000 units |
£434.00
5000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lambda DNA digested with MluI, 0.7% agarose, 7 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
- Superior quality -stringent quality control and industry leading manufacturing process
- Convenient color-coded Five Buffer System
- Includes universal Tango buffer for double-digestions
- BSA premixed in reaction buffers
- Wide selection of restriction endonuclease specificities
Specifications
10 U/μL | |
Mlu I | |
Yes | |
No | |
Traditional Cloning |
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive | |
10X Buffer R | |
37°C | |
Restriction Enzyme |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.