Learn More
Thermo Scientific™ MssI (PmeI) (5 U/μL)
The MssI (PmeI) restriction enzyme recognizes GTTT^AAAC sites and cuts best at 37°C in B buffer (Isoschizomers: PmeI).
Brand: Thermo Scientific™ ER1342
144.62 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Description
Lambda DNA digested with MssI (PmeI), 0.7% agarose, 2 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...G T T T▵A A A C...3'
3'...C A A A▵T T T G...5'
Conditions for 100% Activity:
- 1X Buffer B: 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2 and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- MssI is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Methylation Effects:
- Dam: never overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: may overlap —no effect
- EcoKI: may overlap —blocked
- EcoBI: never overlaps —no effect
Digestion of Agarose-embedded DNA:
- Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours
Specifications
5 U/μL | |
MssI (PmeI) | |
Yes | |
No | |
Restriction Enzyme |
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
10X Buffer B | |
37°C | |
1250 U | |
Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.