Learn More
Thermo Scientific™ NoLimits 15 bp DNA Fragment
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Brand: Thermo Scientific™ SM1381
48.33 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.
Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Highlights
- Chromatography-purified individual DNA fragments
- Broad range: 10 to 20,000 bp
- Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
- Precise DNA concentration
Specifications
Agarose | |
No | |
Sustainable packaging | |
10 μg |
DNA Fragment | |
Agarose Gels | |
NoLimits |
For Research Use Only. Not for use in diagnostic procedures.