Learn More
Thermo Scientific™ NoLimits 900 bp DNA Fragment
NoLimits is a collection of 36 chromatography-purified individual DNA fragments ranging from 10-20,000 bp for applications where a custom DNA size standard is required.
Brand: Thermo Scientific™ SM1491
38.47 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.
Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Specifications
Agarose | |
No | |
Agarose Gels | |
NoLimits |
DNA Fragment | |
10 to 20000 bp | |
Sustainable packaging | |
10 μg |
For Research Use Only. Not for use in diagnostic procedures.