Learn More
Thermo Scientific™ NTP Set, 100 mM Solution
The Thermo Scientific NTP set is a convenient set of 100 mM solutions of each ATP, CTP, GTP and UTP.
Brand: Thermo Scientific™ R0481
73.20 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Description
The Thermo Scientific NTP set is a convenient set of 100 mM solutions of each ATP, CTP, GTP and UTP. These highly stable nucleotides are more than 99% pure and can be used in a variety of molecular biology applications.
Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles
Applications
• aRNA synthesis
• siRNA synthesis
• RNA amplification
• in vitro transcription
Related products
100 mM ATP Solution (Cat. No. R0441)
100 mM CTP Solution (R0451)
100 mM GTP Solution (R0461)
100 mM UTP Solution (R0471)
Specifications
100 mM | |
NTP Set, 100 mM solution (4 x 0.25 mL) Store at –20°C. |
|
Unconjugated | |
NTP Set |
in vitro Transcription, RNA Amplification | |
Dry Ice | |
4 x 0.25 mL | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.