missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Water, nuclease-free

Optimize molecular biology applications with this deionized and 0.22μm membrane-filtered nuclease-free water.

Brand:  Thermo Scientific™ R0581

 View more versions of this product

Product Code. 11893933

  • £21.91 / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Nuclease-free water is deionized and 0.22 μm membrane-filtered nuclease-free water. It is ideal for all .

Ideal for all molecular biology applications.
  • Free of endo-, exodeoxyribonucleases, ribonucleases and phosphatases
TRUSTED_SUSTAINABILITY
Specifications

Specifications

7.6
Vial
4 x 1.25 mL
Molecular Biology
Not DEPC-Treated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.