Learn More
Thermo Scientific™ pBR322 Plasmid DNA
Optimize cloning with Thermo Scientific™ pBR322 DNA, a 4361 bp double-stranded closed circular medium copy plasmid DNA.
Brand: Thermo Scientific™ SD0041
71.73 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 88
Additional Details : Weight : 1.51950kg
Description
pBR322 is one of the most commonly used E.coli cloning vectors.
Highlights
- Isolated from E. coli (dam+, dcm+)
- More than 90% in the supercoiled form
- Purified by chromatography using proprietary patented technology
- For DNA sequence, sequence analysis, and map creation, see the REviewer online free tool.
Specifications
Restriction Enzyme⁄ MCS | |
DNA | |
Cloning | |
100 μg |
0.5 μg/μL | |
DNA | |
Plasmid DNA | |
pBR322 |
For Research Use Only. Not for use in diagnostic procedures.