Learn More
Thermo Scientific™ phiX174 DNA/BsuRI (HaeIII) Marker
Size and approximately quantify small double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels with this conventional marker composed of 11 sharp bands.
£81.00 - £330.00
Specifications
Gel Type | Agarose |
---|---|
Product Type | DNA/BSURI (HAEIII) Marker |
Ready to Load | No |
Gel Compatibility | Agarose Gels |
Green Features | Sustainable packaging |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10570091
|
Thermo Scientific™
SM0251 |
50 μg |
£81.00
50µg |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
11893953
|
Thermo Scientific™
SM0252 |
5 x 50 μg |
£330.00
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
phiX174 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation.
phiX174 DNA/BsuRI (HaeIII) Marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction.
Highlights
- Sizing and approximate quantification of small DNA fragments
- Sharp bands
- Supplied with loading dye for sample DNA
Specifications
Agarose | |
No | |
Sustainable packaging |
DNA/BSURI (HAEIII) Marker | |
Agarose Gels | |
phiX174 DNA/BsuRI (HaeIII) Marker |
For Research Use Only. Not for use in diagnostic procedures.