missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pJET1.2 Forward Sequencing Primer, 23-mer

Thermo Scientific sequencing primers are single-stranded oligonucleotides with 5'-hydroxyl and 3'-hydroxyl ends.

Brand:  Thermo Scientific™ SO501

41.70 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10590091

  • £69.50 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific sequencing primers are single-stranded oligonucleotides with 5'-hydroxyl and 3'-hydroxyl ends. pJET1.2 sequencing primers flank the Eco32I site in the eco47IR gene of positive selection cloning vector pJET1.2. All primers are supplied as 10 µM aqueous solutions.

Applications
• Sequencing of DNA fragments inserted into Eco32I site within the eco47IR gene of pJET1.2p sequence
• Colony screening by PCR

pJET1.2 Primer sequences
• pJET1.2 forward sequencing primer, 23-mer: 5'-d(CGACTCACTATAGGGAGAGCGGC)-3'
• pJET1.2 reverse sequencing primer, 24-mer: 5'-d(AAGAACATCGATTTTCCATGGCAG)-3'

Related products
pJET1.2 Reverse Sequencing Primer, 24-mer (Cat. No. SO511)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Forward Sequencing Primer
Dry Ice
pJET
Sequencing
T3 promoter Sequencing Primer, 24-mer, 10 μM

Store at –20°C.
10 μM
pJET1.2
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.