Learn More
Thermo Scientific™ pUC19 DNA
Optimize cloning with this double-stranded closed circular high copy plasmid DNA, 2686 base pairs with a molecular weight of 1.74 x 106 Da isolated from E.coli.
Brand: Thermo Scientific™ SD0061
75.29 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 88
Additional Details : Weight : 0.00400kg
Description
pUC18 and pUC19 vectors are small, high copy number, E. coli plasmids, 2686 bp in length. They are identical except that they contain multiple cloning sites (MCS) arranged in opposite orientations.
Highlights
- Purified by chromatography using proprietary patented technology
- More than 90% in the supercoiled form
- Isolated from E.coli (dam+, dcm+)
Specifications
Restriction Enzyme⁄ MCS | |
DNA | |
Cloning | |
50 μg |
0.5 μg/μL | |
DNA | |
DNA | |
pUC19 |
For Research Use Only. Not for use in diagnostic procedures.