Learn More
Thermo Scientific™ phiX174 DNA/BsuRI (HaeIII) Marker, ready-to-use
Size and approximately quantify small double-stranded DNA fragments in agarose and non-denaturing polyacrylamide gels. Premixed with Loading Dye for direct loading onto gel.
Brand: Thermo Scientific™ SM0253
137.75 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 88
Additional Details : Weight : 1.51950kg
Description
0.5 µg/lane, 8cm length gel, 1X TBE, 5V/cm, 1.5h. Bands visible: 1353, 1078, 872, 603, 310 281, 271, 234, 194, 118, and 118 bp.
phiX174 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation.
phiX174 DNA/BsuRI (HaeIII) Marker, ready-to-use is premixed with 6X DNA Loading Dye for direct loading onto agarose and polyacrylamide gels.
Highlights
- Sizing and approximate quantification of small DNA fragments
- Sharp bands
- Premixed with 6X DNA Loading Dye for direct loading and room temperature storage
- Supplied with loading dye for sample DNA
Specifications
6X | |
DNA/BSURI (HAEIII) Marker | |
Agarose Gels |
Agarose | |
Yes | |
Sustainable packaging |
For Research Use Only. Not for use in diagnostic procedures.