Learn More
Thermo Scientific™ pUC19 DNA/MspI (HpaII) Marker, ready-to-use
Size and approximately quantify small DNA fragments in agarose and non-denaturing polyacrylamide gels. Premixed with Loading Dye for direct loading onto gel.
Brand: Thermo Scientific™ SM0223
70.78 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 88
Additional Details : Weight : 0.00900kg
Description
0.5 µg/lane, 8 cm length gel, 1X TBE, 5 V/cm, 1.5 h. Bands visible: 501, 489, 404, 331, 242, 190, 147, 111, 110, 67, 34 and 34 bp.
pUC19 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation. pUC19 DNA/MspI (HpaII) Marker, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading onto agarose and polyacrylamide gels.
Highlights
- Sizing and approximate quantification of small DNA fragments
- Sharp bands
- Premixed with 6X DNA Loading Dye for direct loading and room temperature storage
- Supplied with loading dye for sample DNA
Specifications
6X | |
DNA/MSPI (HPAII) Marker | |
Agarose Gels |
Agarose | |
Yes | |
Sustainable packaging |
For Research Use Only. Not for use in diagnostic procedures.