missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RevertAid Reverse Transcriptase (200 U/μL)

Thermo Scientific RevertAid Reverse Transcriptase (RT) is a recombinant M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties.

Brand:  Thermo Scientific™ EP0442

 View more versions of this product

Product Code. 10161310

  • £283.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific RevertAid Reverse Transcriptase (RT) is a recombinant M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity and a RNase H activity specific to RNA in RNA-DNA hybrids that is significantly lower than that of Avian Myeloblastosis Virus (AMV) reverse transcriptase.

Features of RevertAid Reverse Transcriptase include:
• Efficient synthesis of full-length first strand cDNA up to 13 kb
• Optimum activity at 42°C
• Active up to 50°C
• Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, and fluorescein-labeled nucleotides)

Applications
• First strand cDNA synthesis for RT-PCR and real-time RT-PCR
• Synthesis of cDNA for cloning and expression
• Generation of labeled cDNA probes for microarrays
• DNA labeling
• Analysis of RNA by primer extension

Source
RevertAid Reverse Transcriptase is sourced from E.coli cells with a cloned fragment of the pol gene encoding Moloney Murine Leukemia Virus reverse transcriptase.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• RevertAid Reverse Transcriptase (200 U/μL)
• 5X Reaction Buffer

Store at –20°C.

60 min.
42°C
RevertAid
Dry Ice
First-Strand cDNA
Reverse Transcription
RNA
Stand-alone Enzyme
Reverse Transcription
5 x 10,000 Units
Yes
200 U/μL
Separate components
Up to 13 kb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2793214 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0442
Lot Number2799588 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0442
Lot Number2825142 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0442
Lot Number2811358 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0442
Lot Number2849758 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0442
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ RevertAid Reverse Transcriptase (200 U/μL) > 5 x 10,000 Units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.