Learn More
Thermo Scientific™ RNase A, DNase and protease-free (10 mg/mL)
Specifically degrade single-stranded RNA at C and U residues with the endoribonuclease RNase A.
Brand: Thermo Scientific™ EN0531
40.70 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
RNase A, DNase and protease-free is an endoribonuclease that specifically degrades single-stranded RNA at C and U residues.
It cleaves the phosphodiester bond between the 5'-ribose of a nucleotide and the phosphate group attached to the 3'-ribose of an adjacent pyrimidine nucleotide. The resulting 2', 3'-cyclic phosphate is hydrolyzed to the corresponding 3'-nucleoside phosphate (see References 1, 2).
Highlights
- RNase A is free of DNase activity. It is not necessary to heat it before use.

Specifications
10 mg/mL | |
RNase | |
RNase A |
Store at -20°C. | |
10 mg |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.