missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNase I (10 U/μL)

Ribonuclease I (RNase I) is an endoribonuclease that hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates.

Specifications

Concentration 10 U/μL
Enzyme RNase
Product Type RNase I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10568930
View Documents Promotion Details
Thermo Scientific™
EN0601
1000 U N/A
N/A
Edge
10374290
View Documents Promotion Details
Thermo Scientific™
EN0602
5000 U N/A
N/A
Description

Description

Ribonuclease I (RNase I), an endoribonuclease, preferentially hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates via nucleoside 2', 3'-cyclic monophosphate intermediates.

The enzyme does not require any metal ions for activity.

Highlights

  • Stable and active under a wide variety of reaction conditions
  • Can be heat inactivated in 30 minutes at 100°C
Specifications

Specifications

10 U/μL
RNase
RNase I
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.