missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNase T1 (1000 U/μL)

RNase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues.

£63.50 - £262.00

Specifications

Concentration 1000 U/μL
Enzyme RNase
Product Type RNase T1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10314820
View Documents Promotion Details
Thermo Scientific™
EN0541
100,000 U
£63.50
100000 units
Please to purchase this item. Need a web account? Register with us today!
10621621
View Documents Promotion Details
Thermo Scientific™
EN0542
500,000 U
£262.00
500000 units
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific Rnase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues (see Figure 1 in Supporting Data). It cleaves the phosphodiester bond between the 3'-guanylic residue and the 5'-OH residue of adjacent nucleotides with the formation of corresponding intermediate 2', 3'-cyclic phosphate. The reaction products are 3'-GMP and oligonucleotides with a terminal 3'-GMP.

Rnase T1 does not require metal ions for activity.

Applications
  • Removal of RNA from DNA preparations
  • RNA sequencing
  • Ribonuclease protection assays. Used in conjunction with RNase A
  • Removal of RNA from recombinant protein preparations
  • Determination of the level of RNA transcripts synthesized in vitro from DNA templates containing a “G-less cassette”
Specifications

Specifications

1000 U/μL
RNase
RNase T1
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.