missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SatI (Fnu4HI) (10 U/μL)

The SatI (Fnu4HI) restriction enzyme recognizes GC^NGC sites and cuts best at 37°C in G buffer (Isoschizomers: Fnu4HI, Fsp4HI, ItaI).

Brand:  Thermo Scientific™ ER1642

 View more versions of this product

Product Code. 10764501

  • £317.00 / 1000 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Lambda DNA digested with SatI (Fnu4HI), 1.4% agarose, 380 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G C▵N G C...3'

3'...C G N▵C G...5'

Conditions for 100% Activity

  • 1X Buffer G:10mM Tris-HCl (pH7.5 at 37°C), 10mM MgCl2, 50mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer

  • SatI is supplied in:10mM Tris-HCl (pH7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage

  • After 50-fold overdigestion with SatI, more than 60% of the DNA fragments can be ligated in a reaction mixture containing 10-30U of T4 DNA Ligase/1μg of fragments and 10% PEG
  • More than 90% of these can be recut

Methylation Effects

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: may overlap — blocked
  • EcoKI: never overlaps — no effect
  • EcoBI: never overlaps — no effect

Compatible Ends

  • GC^SGC — BcnI, Bme1390I
  • GC^WGC — MvaI, Bme1390I

Note:

At least two copies of SatI (Fnu4HI) recognition site are required for efficient cleavage.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 U/μL
SatI (Fnu4HI)
Yes
No
Restriction Enzyme
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer G
37°C
1,000 units
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.