missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SP6 promoter Sequencing Primer, 18-mer

Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends.

Brand:  Thermo Scientific™ SO116

20.36 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10100950

  • £40.71 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends. The SP6 Promoter Sequencing Primer, 18-mer is complementary to the SP6 RNA Polymerase promoter region and is supplied as 10 μM aqueous solutions.

Applications
• Sequencing of DNA fragments located downstream from the SP6 RNA polymerase promoter sequence in common cloning vectors, such as pTZ19R (Cat. No. SD0141), pTZ57R, and pBluescript II

Promoter Sequence 5'-d (ATTTAGGTGACACTATAG)-3'

Related products
SP6 promoter Sequencing Primer, 24-mer (SO117)
T7 promoter Sequencing Primer, 20-mer (SO118)
T3 promoter Sequencing Primer, 17-mer (SO119)
T3 promoter Sequencing Primer, 24-mer (SO120)

Notes
• Primers cannot be used for certain plasmids that contain truncated, but still fully functional promoters
• Primers are not phosphorylated

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SP6, T3, T7
SP6 promoter Sequencing Primer, 18-mer, 10 μM (56 μL)

Store at –20°C.
10 μM
56 μL
Sequencing
Sequencing Primer
Dry Ice
SP6
pTZ19R, pTZ57R, pBluescript II
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.