Learn More
Thermo Scientific™ SP6 RNA Polymerase, HC (≥100 U/μL)
Thermo Scientific™ SP6 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.
Brand: Thermo Scientific™ EP0133
172.90 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 88
Additional Details : Weight : 0.00900kg
Beschreibung
Thermo Scientific™ Bacteriophage SP6 RNA polymerase is DNA-dependent RNA polymerase with strict specificity for their respective double-stranded promoters.
It catalyzes the 5'=>3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter.- Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
Note:
Consensus promoter sequences: AATTAACCCTCACTAAA G GGAGA - T3 TAATACGACTCACTATA G GGAGA - T7 ATTTAGGTGACACTATA G AAGNG - SP6 The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively (9).
Spezifikation
SP6 RNA Polymerase | |
1000 U |
For Research Use Only. Not for use in diagnostic procedures.