Learn More
Thermo Scientific™ T4 DNA Polymerase (5 U/μL)
Ensure accuracy in DNA synthesis with template-dependent T4 DNA Polymerase that catalyzes the 5'-3' synthesis from primed single-stranded DNA.
Brand: Thermo Scientific™ EP0061
40.72 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 88
Additional Details : Weight : 0.00800kg
Description
T4 DNA Polymerase, a template-dependent DNA polymerase, catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzyme has a 3'-5' exonuclease activity, but lacks 5'-3' exonuclease activity.
Highlights
- Stronger 3'-5' exonuclease activity on single-stranded than on double-stranded DNA and greater (more than 200 times) than DNA polymerase I, E. coli,and Klenow fragment
- Active in Thermo Scientific restriction enzyme, PCR, RT and T4 DNA Ligase buffers
Specifications
T4 DNA Polymerase | |
100 unit | |
1mL of 5X Reaction Buffer |
For Research Use Only. Not for use in diagnostic procedures.