Learn More
Thermo Scientific™ T4 Polynucleotide Kinase (10 U/μL)
T4 Polynucleotide Kinase (T4 PNK) catalyzes the transfer of the gamma-phosphate from ATP to the 5'-OH group of oligonucleotides, ss and dsDNA and RNA.
£43.65 - £189.00
Specifications
Concentration | 10 U/μL |
---|---|
Enzyme | T4 Polynucleotide Kinase |
Compatible Buffer | 10X Reaction Buffer |
Product Type | T4 Polynucleotide Kinase |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10120670
|
Thermo Scientific™
EK0031 |
500 U |
£43.65
500 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10531061
|
Thermo Scientific™
EK0032 |
2500 U |
£189.00
2500 units |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
T4 Polynucleotide Kinase (T4 PNK) catalyzes the transfer of the gamma-phosphate from ATP to the 5'-OH group of single- and double-stranded DNAs and RNAs, oligonucleotides or nucleoside 3'-monophosphates (forward reaction). The reaction is reversible. In the presence of ADP T4 Polynucleotide Kinase exhibits 5'-phosphatase activity and catalyzes the exchange of phosphate groups between 5'-P-oligo-polynucleotides and ATP (exchange reaction).
The enzyme is also a 3'-phosphatase.
Highlights
- Active in Thermo Scientific restriction enzyme, RT, and T4 DNA Ligase buffers
Specifications
10 U/μL | |
T4 Polynucleotide Kinase | |
10X Reaction Buffer | |
T4 Polynucleotide Kinase |
For Research Use Only. Not for use in diagnostic procedures.