Learn More
Thermo Scientific™ T7 RNA Polymerase, HC (200 U/μL)
Thermo Scientific™ T7 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.
Brand: Thermo Scientific™ EP0113
132.60 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Description
Thermo Scientific™ Bacteriophage T7 RNA polymerase is DNA-dependent RNA polymerase with strict specificity its respective double-stranded promoter.
It catalyzes the 5'=>3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter.- Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used:for hybridization, in vitro RNA translation; as aRNA, siRNA, substrate in RNase protection assays, template for genomic DNA sequencing.
Note:
Consensus promoter sequences: TAATACGACTCACTATA G GGAGA. The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively.

Specifications
T7 RNA Polymerase | |
25,000 units | |
>200U/µL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.