Learn More
Thermo Scientific™ T7 RNA Polymerase (20 U/μL)
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Bacteriophage T7 RNA polymerase is DNA-dependent RNA polymerase with strict specificity its respective double-stranded promoter.
- Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used:for hybridization, in vitro RNA translation; as aRNA, siRNA, substrate in RNase protection assays, template for genomic DNA sequencing.
Note:
Consensus promoter sequences: TAATACGACTCACTATA G GGAGA. The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively.
Specifications
Specifications
| Polymerase | T7 RNA Polymerase |
| Quantity | 5 x 5,000 units |
| Concentration | 20U/µL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.