Learn More
Thermo Scientific™ TopVision Agarose
TopVision Agarose is highly purified agarose suitable for analytical and preparative electrophoresis of nucleic acids.
Brand: Thermo Scientific™ R0492
330.31 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific TopVision Agarose provides optimal concentration between 0.7 to 2% in all typical buffer systems. It is a highly purified agarose with very low EEO values certified by strict quality control test procedures.
Specifications
Agarose Gel | |
88 °C | |
500 g |
100 to >30000 bp | |
TopVision | |
Gel Electrophoresis |