Learn More
Thermo Scientific™ TopVision Low Melting Point Agarose
Perform in-gel enzymatic process experiments and nucleic acid recovery after electrophoresis with Thermo Scientific™ TopVision™ Low Melting Point Agarose.
Brand: Thermo Scientific™ R0801
119.70 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 2.44490kg
Description
Top Vision Low Melting Point Agarose is highly purified agarose with a low melting temperature certified by strict quality control test procedures. It is used for in-gel enzymatic process experiments and for nucleic acids recovery from gels using Agarase (#EO0461).
Highlights
- Optimal concentration between 0.7-2% in all typical buffer systems
- GQ (Genetic Quality) certified, which ensures that nucleic acids recovered from preparative gels can be used for downstream applications (enzymatic reactions etc.)
- Low DNA/RNA binding
- Excellent gel transparency
- DNase and RNase free
- Suitable for RNA analysis
Specifications
Agarose Gel | |
65.5 °C | |
Gel Electrophoresis |
100 to >30000 bp | |
TopVision |
For Research Use Only. Not for use in diagnostic procedures.