missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ XmaJI (AvrII) (10 U/μL)

Cut at C^CTAGG sites with XmaJI (AvrII) restriction enzyme, which functions best at 37°C in Tango buffer (Isoschizomers: AspA2I, AvrII, BlnI).

Brand:  Thermo Scientific™ ER1561

146.81 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 10470090

  • £160.00 / 200 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Lambda DNA digested with XmaJI, 1% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...C▵C T A G G...3'

3'...G G A T C▵C...5'

Conditions for 100% Activity

  • 1X Buffer Tango:33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer

  • XmaJI is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage

  • After 50-fold overdigestion with XmaJI, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects

  • Dam: never overlaps-no effect
  • Dcm: never overlaps-no effect
  • CpG: never overlaps-no effect
  • EcoKI: never overlaps-no effect
  • EcoBI: never overlaps-no effect

Digestion of Agarose-embedded DNA

  • Minimum 10U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Compatible Ends

  • BcuI, NheI, XbaI, Eco130I (C▴CTAGG)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 U/μL
XmaJI (AvrII)
Yes
No
Restriction Enzyme
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
10X Buffer Tango
37°C
200 U
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.