Learn More
Thermo Scientific™ Pyrophosphatase, inorganic (0.1 U/μL)
Catalyze the hydrolysis of inorganic pyrophosphate to two orthophosphates.
Brand: Thermo Scientific™ EF0221
36.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 88
Additional Details : Weight : 1.51950kg
Description
Thermo Scientific Pyrophosphatase, Inorganic catalyzes the hydrolysis of inorganic pyrophosphate into two orthophosphates (see Figure 1 in Supporting data). The enzyme requires a divalent metal cation with Mg2+ conferring the highest activity.
Highlights
- Active in Thermo Scientific buffers for DNA polymerases, RNA polymerases, and reverse transcriptases
Specifications
0.1 U/μL | |
Storage (Dilution) Buffer | |
Pyrophosphatase |
Pyrophosphatase | |
10 U |
For Research Use Only. Not for use in diagnostic procedures.