Learn More
Ferric Ammonium Sulphate R2 Solution According to European Pharmacopoeia (EP), Reagecon
BIOPHARMA
High-purity, stable product developed and tested for European pharmacopoeia (EP) applications including the preparation of other reagents (4.1.1), the manufacture of containers (3.1), and as a reagent in the performance of tests outlined in several EP monographs.
Brand:  Reagecon 1037702
Product Code. 11404689
  • £143.00 / 1 L
Explore more special offers
Description
Description
  • European Pharmacopoeia Reagent, Solution
  • High-purity stable product developed and tested for European pharmacopoeia (EP) applications.
  • Used for the preparation of other reagents (4.1.1), manufacture of containers (3.1), and as a reagent in the performance of tests outlined in several EP monographs
  • This Reagecon EP reagent is accurate, stable and reproducible.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
Specifications
Specifications
HDPE Bottle GL45
1 L
EP Testing