missing translation for 'onlineSavingsMsg'
Learn More

Ferrocenecarboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11303658
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Ferrocenecarboxaldehyde, is used to prepare chiral ferrocene aziridinylmethanols for selective azomethine ylide cycloaddtion. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12093-10-6
Molecular Formula C11H10FeO
Molecular Weight (g/mol) 214.05
MDL Number MFCD00001429
InChI Key UQTCQJVPLIVCAX-UHFFFAOYNA-N
Synonym [Cyclopentadienyl(formylcyclopentadienyl)iron]
IUPAC Name (cyclopenta-2,4-dien-1-yl)(2-formylcyclopenta-2,4-dien-1-yl)iron
SMILES [Fe].c1cccc1.O=Cc1cccc1

Specifications

Melting Point 124°C
Quantity 2 g
Sensitivity Air sensitive
Solubility Information It is soluble in water.
Formula Weight 214.05
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Ferrocenecarboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.