Learn More
Corning Media Tech Fetal Bovine Serum
Supplement for cell culture.
Brand: Corning Media Tech 35-079-CV
Code : NEW
Additional Details : Weight : 0.54000kg
Description
- Raw pooled serum is filtered through series of 0.1μ sterilizing grade filters
- Low endotoxin levels
- Low hemoglobin levels
Specifications
Liquid | |
500 mL | |
Sterile-filtered | |
Cell and tissue culture |
Fetal Bovine Serum | |
Sterile | |
<20 EU/mL | |
<30 mg/dL |