missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, dialyzed, US origin
GREENER_CHOICE

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

Brand:  Gibco™ 26400044

677.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11520646

  • £774.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Fetal Bovine Serum
Dialyzed
Frozen
Fetal
United States
Bottle
500 mL
Sustainable packaging
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.