missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, dialyzed, US origin
GREENER_CHOICE

Product Code. 15605639
Click to view available options
Quantity:
50 mL
500 mL
Format:
Bottle
One Shot
Unit Size:
500mL
50mL
This item is not returnable. View return policy

Product Code. 15605639

Brand: Gibco™ A3382001

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

Gibco Fetal bovine sera include a broad range of specialty sera, earning the trust of researchers with consistent quality and award-winning support that helps meet your unique research requirements. Dialyzed FBS provides a more controlled and defined environment with the removal of small molecules, enhancing outcome accuracy and reliability.

  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (specification is checked and recorded; levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Bovine
Age Fetal
Country of Origin United States
Form Liquid
Format One Shot
Product Type Fetal Bovine Serum
Quantity 50 mL
Serum Treatment Dialyzed
Sterility Sterile
Product Line One Shot
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.