missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, exosome-depleted
GREENER_CHOICE

Gibco™ Exosome-Depleted FBS streamlines and standardizes cell culture workflows for exosome research. It is ready to use and performance tested to help save time and enable optimal exosome preparations.

Brand:  Gibco™ A2720801

 View more versions of this product

Product Code. 15434573

  • £1258.00 / 500mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Gibco™ Exosome-Depleted FBS streamlines and standardizes cell culture workflows for exosome research. It is ready to use and performance tested to help save time and enable optimal exosome preparations.

  • Lowest exosome background
  • Enables consistent results: each lot tested for cell culture performance
  • No preparation needed: ultracentrifugation is not required

Exosome-Depleted FBS has been depleted of ≥90% of endogenous exosomes and is intended for use in cell culture exosome isolation assays. Every lot is tested for exosome depletion and culture performance, and our standard quality control assays are conducted to help ensure optimal and consistent results. This ready-to-use FBS for cell culture exosome isolations helps eliminate the risks associated with FBS reagent preparation by ultracentrifugation, for optimal assay results and increased experimental throughput.

Recommended Storage

Can be thawed and refrozen

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
United States
Bottle
500 mL
Sterile
Frozen
Fetal
Liquid
Fetal Bovine Serum
Exosome-depleted
Sustainable packaging
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.