Läs mer
Gibco™ Fetal Bovine Serum, exosome-depleted

Beskrivning
Gibco™ Exosome-Depleted FBS streamLines and standardizes cell culture workflows for exosome research. It is ready to use and performance tested to help save time and enable optimal exosome preparations.
- Lowest exosome background
- Enables consistent results: each lot tested for cell culture performance
- No preparation needed: ultracentrifugation is not required
Exosome-Depleted FBS has been depleted of ≥90% of endogenous exosomes and is intended for use in cell culture exosome isolation assays. Every lot is tested for exosome depletion and culture performance, and our standard quality control assays are conducted to help ensure optimal and consistent results. This ready-to-use FBS for cell culture exosome isolations helps eliminate the risks associated with FBS reagent preparation by ultracentrifugation, for optimal assay results and increased experimental throughput.
Specifikationer
Specifikationer
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Bovine |
| Age | Fetal |
| Country of Origin | United States |
| Form | Liquid |
| Format | One Shot |
| Product Type | Fetal Bovine Serum |
| Quantity | 50 mL |
| Serum Treatment | Exosome-depleted |
| Sterility | Sterile |
| Visa mer |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.