missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Fetal Bovine Serum, Premium
Click to view available options
Quantity:
50 mL
500 mL
10 x 50 mL
Format:
Bottle
One Shot
Heat Inactivated:
No
Yes
Gamma-irradiated:
No
Yes
Unit Size:
500mL
50mL
Pack of 10
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Sera category: Premium
- Use for general cell culture with common cell lines
- Low endotoxin and hemoglobin levels
- Endotoxin level: <10 EU/mL
- Hemoglobin level: <25 mg/dL
- Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
- US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco serum delivers
- ISO 13485–certified; processed in FDA registered facilities
- Triple-filtered at 0.1 μm
- Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Specifications
Specifications
| Content And Storage | Store at ≤-10°C. |
| Species | Cattle/Bovine |
| Age | Fetal |
| Country of Origin | Refer to CoA for specific origin |
| Form | Frozen |
| Format | One Shot |
| Product Type | Fetal Bovine Serum |
| Quantity | 50 mL |
| Sterility | Sterile-filtered |
| Sterilization Method | Triple-filtered, 0.1 μm |
| Show More |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction