Learn More
Gibco™ Fetal Bovine Serum (FBS), Premium, heat-inactivated
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
£63.50 - £592.00
Specifications
Content And Storage | Store at ≤-10°C. |
---|---|
Age | Fetal |
Species | Cattle/Bovine |
Country of Origin | Refer to CoA for specific origin |
Form | Frozen |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
17934731
|
Gibco™
A5670501 |
50 mL | One Shot |
£63.50
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17924671
|
Gibco™
A5670502 |
10 x 50 mL | One Shot |
£592.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17914671
|
Gibco™
A5670801 |
500 mL | Bottle |
£512.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
Gibco sera: trusted quality for consistent performance
Sera category: Premium
- Use for general cell culture with common cell lines
- Low endotoxin and hemoglobin levels
- Endotoxin level: <10 EU/mL
- Hemoglobin level: <25 mg/dL
- Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
- US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco serum delivers
- ISO 13485–certified; processed in FDA registered facilities
- Triple-filtered at 0.1 μm
- Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Specifications
Store at ≤-10°C. | |
Cattle/Bovine | |
Frozen | |
Heat-inactivated | |
Triple-filtered, 0.1 μm | |
Up to 60 quality tests including 9CFR virus testing, endotoxin, and performance | |
Less waste | |
Premium |
Fetal | |
Refer to CoA for specific origin | |
Fetal Bovine Serum | |
Sterile-filtered | |
Premium | |
≤10 EU/mL | |
≤25 mg/dL | |
Dry Ice |
Safety and Handling
missing translation for 'shelfLife' : 5 Years
Your input is important to us. Please complete this form to provide feedback related to the content on this product.